4-Brom-3-chlorbenzhydrazid

Lieferant: FLUOROCHEM

018253-1G
FLUO018253-1GEA 159 EUR
FLUO018253-1G
4-Brom-3-chlorbenzhydrazid
4-Brom-3-chlorbenzhydrazid
Formel: C₇H₆BrClN₂O
MDL: MFCD00270096
CAS-Nummer: 148993-19-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR