3-Brom-2,4-dichlortoluol

Lieferant: FLUOROCHEM

018267-5G
FLUO018267-5GEA 575 EUR
FLUO018267-5G
3-Brom-2,4-dichlortoluol
3-Brom-2,4-dichlortoluol
Formel: C₇H₅BrCl₂
MDL: MFCD00079712
CAS-Nummer: 206559-41-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR