(2-Brom-4,5-dimethoxyphenyl)acetonitril

Lieferant: FLUOROCHEM

Synonyme: 2-(2-Bromo-4,5-dimethoxyphenyl)acetonitrile

018271-1G 018271-5G
FLUO018271-1GEA 39.9 EUR
FLUO018271-1G FLUO018271-5G
(2-Brom-4,5-dimethoxyphenyl)acetonitril
(2-Brom-4,5-dimethoxyphenyl)acetonitril
Formel: C₁₀H₁₀BrNO₂
Molecular Weight: 256.1000 g/mol
MDL: MFCD00016385
CAS-Nummer: 51655-39-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR