4-Brom-2,6-dimethylphenylisothiocyanat

Lieferant: FLUOROCHEM

018272-1G
FLUO018272-1GEA 60.9 EUR
FLUO018272-1G
4-Brom-2,6-dimethylphenylisothiocyanat
4-Brom-2,6-dimethylphenylisothiocyanat
Formel: C₉H₈BrNS
MDL: MFCD00041088
CAS-Nummer: 32265-82-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR