4-Bromdiphenylmethan

Lieferant: FLUOROCHEM

Synonyme: 1-Benzyl-4-bromobenzene

018275-5G 018275-1G
FLUO018275-5GEA 269 EUR
FLUO018275-5G FLUO018275-1G
4-Bromdiphenylmethan
4-Bromdiphenylmethan
Formel: C₁₃H₁₁Br
Molecular Weight: 247.130 g/mol
MDL: MFCD00053165
CAS-Nummer: 2116-36-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR