N-(6-Bromhexyl)phthalimid
Lieferant: FLUOROCHEM
018278-5G
FLUO018278-5GEA
102
EUR
FLUO018278-5G
N-(6-Bromhexyl)phthalimid
N-(6-Bromhexyl)phthalimid
Formel:
C₁₄H₁₆BrNO₂ Molecular Weight: 310,19 g/mol |
MDL:
MFCD00023098 CAS-Nummer: 24566-79-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...