2-Brom-5-iodtoluol
Lieferant: FLUOROCHEM
|
Achtung
|
018282-25G
FLUO018282-25GEA
84.9
EUR
FLUO018282-25G
2-Brom-5-iodtoluol
2-Brom-5-iodtoluol
Formel:
C₇H₆BrI Molecular Weight: 296,93 g/mol |
MDL:
MFCD00070749 CAS-Nummer: 202865-85-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...