5-Brom-2-iodtoluol

Lieferant: FLUOROCHEM

018284-10G
FLUO018284-10GEA 42.9 EUR
FLUO018284-10G
5-Brom-2-iodtoluol
5-Brom-2-iodtoluol
Formel: C₇H₆BrI
MDL: MFCD00060664
CAS-Nummer: 116632-39-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR