5-Bromisatosäureanhydrid
Lieferant: FLUOROCHEM
018286-10G
FLUO018286-10GEA
0
EUR
FLUO018286-10G
5-Bromisatosäureanhydrid
5-Bromisatosäureanhydrid
Formel:
C₈H₄BrNO₃ Molecular Weight: 242,03 g/mol |
MDL:
MFCD00016921 CAS-Nummer: 4692-98-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...