2-Brom-4-isopropylanilin

Lieferant: FLUOROCHEM

018287-5G
FLUO018287-5GEA 0 EUR
FLUO018287-5G
2-Brom-4-isopropylanilin
2-Brom-4-isopropylanilin
Formel: C₉H₁₂BrN
Molecular Weight: 214,1 g/mol
Siedepunkt: 139…141 °C (20 mmHg)
Dichte: 1,35 g/cm³ (20 °C)
MDL: MFCD00051585
CAS-Nummer: 51605-97-1
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR