(3-Brom-4-methoxyphenyl)acetonitril

Lieferant: FLUOROCHEM

Synonyme: 2-(3-Bromo-4-methoxyphenyl)acetonitrile

018290-10G
FLUO018290-10GEA 0 EUR
FLUO018290-10G
(3-Brom-4-methoxyphenyl)acetonitril
(3-Brom-4-methoxyphenyl)acetonitril
Formel: C₉H₈BrNO
Molecular Weight: 226,08 g/mol
MDL: MFCD00016391
CAS-Nummer: 772-59-8
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR