2-Brom-4-methylanisol

Lieferant: FLUOROCHEM
Achtung

Synonyme: 3-Brom-4-methoxytoluol, 1-Brom-2-methoxy-5-methylbenzol

018292-10G
FLUO018292-10GEA 0 EUR
FLUO018292-10G
2-Brom-4-methylanisol
2-Brom-4-methylanisol
Formel: C₈H₉BrO
Molecular Weight: 201,06 g/mol
MDL: MFCD00017783
CAS-Nummer: 22002-45-5
EINECS: 244-705-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR