2-Brom-p-toluylsäure

Lieferant: FLUOROCHEM

018293-5G
FLUO018293-5GEA 48.8 EUR
FLUO018293-5G
2-Brom-p-toluylsäure
2-Brom-p-toluylsäure
Formel: C₈H₇BrO₂
Molecular Weight: 215.05 g/mol
MDL: MFCD00045797
CAS-Nummer: 7697-27-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR