3-Brom-o-toluylsäure

Lieferant: FLUOROCHEM

018295-5G
FLUO018295-5GEA 0 EUR
FLUO018295-5G
3-Brom-o-toluylsäure
3-Brom-o-toluylsäure
Formel: C₈H₇BrO₂
Molecular Weight: 215,05 g/mol
MDL: MFCD00270097
CAS-Nummer: 76006-33-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR