4-Brom-m-toluylsäure

Lieferant: FLUOROCHEM
Achtung

018296-10G
FLUO018296-10GEA 57.9 EUR
FLUO018296-10G
4-Brom-m-toluylsäure
4-Brom-m-toluylsäure
Formel: C₈H₇BrO₂
MDL: MFCD00039526
CAS-Nummer: 7697-28-1
EINECS: 231-713-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR