2-Brom-4-methyl-6-nitrophenol

Lieferant: FLUOROCHEM

018298-5G
FLUO018298-5GEA 0 EUR
FLUO018298-5G
2-Brom-4-methyl-6-nitrophenol
2-Brom-4-methyl-6-nitrophenol
Formel: C₇H₆BrNO₃
Molecular Weight: 232,0300 g/mol
MDL: MFCD00060536
CAS-Nummer: 20039-91-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR