4-Brom-o-kresol

Lieferant: FLUOROCHEM
Achtung

Synonyme: 4-Brom-2-methylphenol, 5-Brom-2-hydroxytoluol, 1-Brom-4-hydroxy-3-methylbenzol

018299-5G
FLUO018299-5GEA 48.8 EUR
FLUO018299-5G
4-Brom-o-kresol
4-Brom-o-kresol
Formel: C₇H₇BrO
Molecular Weight: 187,04 g/mol
MDL: MFCD00055435
CAS-Nummer: 2362-12-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR