4-Brom-m-kresol
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
4-Brom-3-methylphenol, 2-Brom-5-hydroxytoluol, 1-Brom-4-hydroxy-2-methylbenzol
018300-10G
FLUO018300-10GEA
36.9
EUR
FLUO018300-10G
4-Brom-m-kresol
4-Brom-m-kresol
Formel:
C₇H₇BrO Molecular Weight: 187,04 g/mol Siedepunkt: 142…145 °C (23 mmHg) Schmelzpunkt: 63 °C |
MDL:
MFCD00079723 CAS-Nummer: 14472-14-1 EINECS: 238-464-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...