4-Brom-2-nitroanisol

Lieferant: FLUOROCHEM

Synonyme: 4-Bromo-1-methoxy-2-nitrobenzene

018301-5G
FLUO018301-5GEA 33.8 EUR
FLUO018301-5G
4-Brom-2-nitroanisol
4-Brom-2-nitroanisol
Formel: C₇H₆BrNO₃
MDL: MFCD00055529
CAS-Nummer: 33696-00-3
EINECS: 251-642-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR