N-(5-Brompentyl)phthalimid

Lieferant: FLUOROCHEM

018303-10G
FLUO018303-10GEA 116 EUR
FLUO018303-10G
N-(5-Brompentyl)phthalimid
N-(5-Brompentyl)phthalimid
Formel: C₁₃H₁₄BrNO₂
MDL: MFCD00060522
CAS-Nummer: 954-81-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR