3-(3-Bromphenyl)propionsäure
Lieferant: FLUOROCHEM
018311-5G
FLUO018311-5GEA
60.5
EUR
FLUO018311-5G
3-(3-Bromphenyl)propionsäure
3-(3-Bromphenyl)propionsäure
Formel:
C₉H₉BrO₂ |
MDL:
MFCD01310792 CAS-Nummer: 42287-90-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...