3-(4-Bromphenyl)propionsäure

Lieferant: FLUOROCHEM

018312-1G 018312-5G
FLUO018312-1GEA 31.1 EUR
FLUO018312-1G FLUO018312-5G
3-(4-Bromphenyl)propionsäure
3-(4-Bromphenyl)propionsäure
Formel: C₉H₉BrO₂
MDL: MFCD01310793
CAS-Nummer: 1643-30-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR