3-(3-Bromphenyl)propiononitril
Lieferant: FLUOROCHEM
018313-5G
FLUO018313-5GEA
347
EUR
FLUO018313-5G
3-(3-Bromphenyl)propiononitril
3-(3-Bromphenyl)propiononitril
Formel:
C₉H₈BrN |
MDL:
MFCD01310794 CAS-Nummer: 376646-63-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...