4'-Butylacetophenon
Lieferant: FLUOROCHEM
Synonyme:
1-(4-Butylphenyl)ethanon, 4-Butyl-1-acetylbenzol, p-Butylacetophenon
018331-10G
FLUO018331-10GEA
0
EUR
FLUO018331-10G
4'-Butylacetophenon
4'-Butylacetophenon
Formel:
C₁₂H₁₆O Molecular Weight: 176,26 g/mol |
MDL:
MFCD00017500 CAS-Nummer: 37920-25-5 EINECS: 253-715-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...