3-(Butylamino)propionitril

Lieferant: FLUOROCHEM

018332-10G
FLUO018332-10GEA 0 EUR
FLUO018332-10G
3-(Butylamino)propionitril
3-(Butylamino)propionitril
Formel: C₇H₁₄N₂
MDL: MFCD00019853
CAS-Nummer: 693-51-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR