3-(tert-Butylamino)propionitril

Lieferant: FLUOROCHEM

Synonyme: 3-(tert-Butylamino)propanenitrile

018333-10G
FLUO018333-10GEA 0 EUR
FLUO018333-10G
3-(tert-Butylamino)propionitril
3-(tert-Butylamino)propionitril
Formel: C₇H₁₄N₂
Molecular Weight: 126,1995 g/mol
MDL: MFCD00045637
CAS-Nummer: 21539-53-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR