4-tert-Butylbenzhydrazid

Lieferant: FLUOROCHEM

Synonyme: 4-tert-Butylbenzol-1-carbohydrazid, 4-tert-Butylbenzoesäurehydrazid, 4-tert-Butylbenzohydrazid

CFF582.2.5 CFF582.5 018335-10G
CHMPCFF582.2.5EA 401 EUR
CHMPCFF582.2.5 CHMPCFF582.5 FLUO018335-10G
4-tert-Butylbenzhydrazid
4-tert-Butylbenzhydrazid
Formel: C₁₁H₁₆N₂O
Molecular Weight: 192,26 g/mol
MDL: MFCD00014763
CAS-Nummer: 43100-38-5
EINECS: 256-090-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR