4-tert-Butylbenzonitril
Lieferant: FLUOROCHEM
Synonyme:
p-tert-Butylbenzonitril
018336-5G
FLUO018336-5GEA
34.1
EUR
FLUO018336-5G
4-tert-Butylbenzonitril
4-tert-Butylbenzonitril
Formel:
C₁₁H₁₃N Molecular Weight: 159,23 g/mol Siedepunkt: 67…69 °C (0,1 mmHg) Dichte: 0,9492 g/cm³ (20 °C) |
MDL:
MFCD00075840 CAS-Nummer: 4210-32-6 EINECS: 224-137-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...