1-tert-Butyl-2-thioharnstoff

Lieferant: FLUOROCHEM

018344-5G
FLUO018344-5GEA 315 EUR
FLUO018344-5G
1-tert-Butyl-2-thioharnstoff
1-tert-Butyl-2-thioharnstoff
Formel: C₅H₁₂N₂S
MDL: MFCD00041192
CAS-Nummer: 7204-48-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR