4-Chlorbenzamidin Hydroiodid

Lieferant: FLUOROCHEM

Synonyme: 4-Chlorbenzoesäureamidin Hydroiodid, 4-Chlorphenylamidin Hydroiodid

018357-5G
FLUO018357-5GEA 323 EUR
FLUO018357-5G
4-Chlorbenzamidin Hydroiodid
4-Chlorbenzamidin Hydroiodid
Formel: C₇H₈ClIN₂
Molecular Weight: 282,51 g/mol
MDL: MFCD00051987
CAS-Nummer: 115297-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR