2-Chlorbenzhydrazid
Lieferant: FLUOROCHEM
Synonyme:
2-Chlorobenzoic hydrazide
018358-10G
FLUO018358-10GEA
93.8
EUR
FLUO018358-10G
2-Chlorbenzhydrazid
2-Chlorbenzhydrazid
Formel:
C₇H₇ClN₂O Molecular Weight: 170,6 g/mol |
MDL:
MFCD00007597 CAS-Nummer: 5814-05-1 EINECS: 227-380-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...