3-Chlorbenzhydrazid

Lieferant: FLUOROCHEM

Synonyme: 3-Chlorobenzoic hydrazide

018359-10G
FLUO018359-10GEA 85.1 EUR
FLUO018359-10G
3-Chlorbenzhydrazid
3-Chlorbenzhydrazid
Formel: C₇H₇ClN₂O
Molecular Weight: 170,6 g/mol
MDL: MFCD00014757
CAS-Nummer: 1673-47-8
EINECS: 216-812-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR