4-Chlorbenzhydrazid

Lieferant: FLUOROCHEM

Synonyme: 4-Chlorobenzoic hydrazide

018360-10G
FLUO018360-10GEA 0 EUR
FLUO018360-10G
4-Chlorbenzhydrazid
4-Chlorbenzhydrazid
Formel: C₇H₇ClN₂O
Molecular Weight: 170,6 g/mol
Siedepunkt: 354 °C (1013 hPa)
Schmelzpunkt: 164 °C
MDL: MFCD00007603
CAS-Nummer: 536-40-3
EINECS: 208-632-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR