5-Chlor-2-benzofurancarbonsäure

Lieferant: FLUOROCHEM

Synonyme: 5-Chlorobenzo[b]furan-2-carboxylic acid

018361-1G
FLUO018361-1GEA 60.9 EUR
FLUO018361-1G
5-Chlor-2-benzofurancarbonsäure
5-Chlorobenzofuran-2-carboxylic acid
Formel: C₉H₅ClO₃
Molecular Weight: 196,59 g/mol
MDL: MFCD00060512
CAS-Nummer: 10242-10-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR