3-Chlor-2,6-dibrom-4-methylanilin

Lieferant: FLUOROCHEM

Synonyme: 3-Chlor-2,6-dibrom-p-toluidin

018364-5G 018364-25G
FLUO018364-5GEA 43.1 EUR
FLUO018364-5G FLUO018364-25G
3-Chlor-2,6-dibrom-4-methylanilin
3-Chlor-2,6-dibrom-4-methylanilin
Formel: C₇H₆Br₂ClN
MDL: MFCD00051580
CAS-Nummer: 84483-22-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR