1-Chlor-2,4-dimethoxybenzol
Lieferant: FLUOROCHEM
018365-10G
FLUO018365-10GEA
395
EUR
FLUO018365-10G
1-Chlor-2,4-dimethoxybenzol
1-Chlor-2,4-dimethoxybenzol
Formel:
C₈H₉ClO₂ Molecular Weight: 172,61 g/mol |
MDL:
MFCD00060636 CAS-Nummer: 7051-13-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...