4-Chlor-3,5-dinitrotoluol

Lieferant: FLUOROCHEM

018368-10G
FLUO018368-10GEA 0 EUR
FLUO018368-10G
4-Chlor-3,5-dinitrotoluol
4-Chlor-3,5-dinitrotoluol
Formel: C₇H₅ClN₂O₄
MDL: MFCD00060535
CAS-Nummer: 5264-65-3
EINECS: 226-080-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR