4'-(2-Chlorethyl)acetophenon
Lieferant: FLUOROCHEM
Synonyme:
1-[4-(2-Chloroethyl)phenyl]ethan-1-one
018371-10G
FLUO018371-10GEA
312
EUR
FLUO018371-10G
4'-(2-Chlorethyl)acetophenon
4'-(2-Chlorethyl)acetophenon
Formel:
C₁₀H₁₁ClO Molecular Weight: 182,65 g/mol |
MDL:
MFCD00016350 CAS-Nummer: 69614-95-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...