1-Chlor-2-fluor-4-iodbenzol

Lieferant: FLUOROCHEM

Synonyme: 4-Chloro-3-fluoroiodobenzene

018374-5G
FLUO018374-5GEA 36.9 EUR
FLUO018374-5G
1-Chlor-2-fluor-4-iodbenzol
1-Chloro-2-fluoro-4-iodobenzene
Formel: C₆H₃ClFI
Molecular Weight: 256,4400 g/mol
MDL: MFCD00042580
CAS-Nummer: 202982-67-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR