4-Chlorsalicylsäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: 4-Chloro-2-hydroxybenzoic acid

018377-10G
FLUO018377-10GEA 0 EUR
FLUO018377-10G
4-Chlorsalicylsäure
4-Chlorsalicylsäure
Formel: C₇H₅ClO₃
Molecular Weight: 172,57 g/mol
MDL: MFCD00002449
CAS-Nummer: 5106-98-9
EINECS: 225-829-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR