4-Chlor-2-iodtoluol

Lieferant: FLUOROCHEM

Synonyme: 1-Chlor-3-iod-4-methylbenzol

018385-10G
FLUO018385-10GEA 36.9 EUR
FLUO018385-10G
4-Chlor-2-iodtoluol
4-Chlor-2-iodtoluol
Formel: C₇H₆ClI
Molecular Weight: 252,48 g/mol
MDL: MFCD00060661
CAS-Nummer: 33184-48-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR