5-Chlor-2-iodtoluol

Lieferant: FLUOROCHEM

018386-5G
FLUO018386-5GEA 0 EUR
FLUO018386-5G
5-Chlor-2-iodtoluol
5-Chlor-2-iodtoluol
Formel: C₇H₆ClI
Molecular Weight: 252,48 g/mol
MDL: MFCD00060665
CAS-Nummer: 23399-70-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR