2-Chlormesitylen

Lieferant: FLUOROCHEM

Synonyme: 2,4,6-Trimethylchlorbenzol

018387-10G
FLUO018387-10GEA 311 EUR
FLUO018387-10G
2-Chlormesitylen
2-Chlormesitylen
Formel: C₉H₁₁Cl
Molecular Weight: 154,64 g/mol
MDL: MFCD00045173
CAS-Nummer: 1667-04-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR