3-Chlor-4-methoxybenzoesäure

Lieferant: FLUOROCHEM

Synonyme: 3-Chlor-p-anissäure

018388-5G
FLUO018388-5GEA 69.3 EUR
FLUO018388-5G
3-Chlor-4-methoxybenzoesäure
3-Chlor-4-methoxybenzoesäure
Formel: C₈H₇ClO₃
Molecular Weight: 186,59 g/mol
MDL: MFCD00016512
CAS-Nummer: 37908-96-6
EINECS: 253-708-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR