2-Chlor-4-methylanisol

Lieferant: FLUOROCHEM

Synonyme: 2-Chloro-1-methoxy-4-methylbenzene, 3-Chloro-4-methoxytoluene

018390-10G
FLUO018390-10GEA 282 EUR
FLUO018390-10G
2-Chlor-4-methylanisol
2-Chlor-4-methylanisol
Formel: C₈H₉ClO
Molecular Weight: 156,609 g/mol
MDL: MFCD00060649
CAS-Nummer: 22002-44-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR