4-Chlor-2-methylanisol

Lieferant: FLUOROCHEM

Synonyme: 5-Chlor-2-methoxytoluol, 1-Chlor-4-methoxy-3-methylbenzol

018391-10G
FLUO018391-10GEA 143 EUR
FLUO018391-10G
4-Chlor-2-methylanisol
4-Chlor-2-methylanisol
Formel: C₈H₉ClO
Molecular Weight: 156,61 g/mol
Siedepunkt: 212…214 °C (1013 hPa)
Schmelzpunkt: 37 °C
MDL: MFCD00070771
CAS-Nummer: 3260-85-3
EINECS: 221-860-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR