4-Chlor-o-toluylsäure

Lieferant: FLUOROCHEM
Achtung

018393-1G
FLUO018393-1GEA 0 EUR
FLUO018393-1G
4-Chlor-o-toluylsäure
4-Chlor-o-toluylsäure
Formel: C₈H₇ClO₂
Molecular Weight: 170,6 g/mol
MDL: MFCD00045852
CAS-Nummer: 7499-07-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR