3-Chlor-2-methylbenzylamin

Lieferant: FLUOROCHEM

018396-1G
FLUO018396-1GEA 55.8 EUR
FLUO018396-1G
3-Chlor-2-methylbenzylamin
3-Chlor-2-methylbenzylamin
Formel: C₈H₁₀ClN
MDL: MFCD00053644
CAS-Nummer: 226565-61-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR