4-Chlor-2-methylbenzylamin

Lieferant: FLUOROCHEM

018397-1G
FLUO018397-1GEA 76.2 EUR
FLUO018397-1G
4-Chlor-2-methylbenzylamin
4-Chlor-2-methylbenzylamin
Formel: C₈H₁₀ClN
MDL: MFCD00047932
CAS-Nummer: 27917-11-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR