5-Chlor-2-methylbenzylamin

Lieferant: FLUOROCHEM

018398-1G
FLUO018398-1GEA 69.3 EUR
FLUO018398-1G
5-Chlor-2-methylbenzylamin
5-Chlor-2-methylbenzylamin
Formel: C₈H₁₀ClN
Molecular Weight: 155,625 g/mol
MDL: MFCD01310800
CAS-Nummer: 27917-13-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR